Fmoc-His(Boc)-OPfp

Supplier: BACHEM AMERICAS INC

4015229.0001 4015229.0025
B-2000.0001BAEA 232.35 CAD
B-2000.0001BA B-2000.0025BA
Fmoc-His(Boc)-OPfp
Fmoc-His(Boc)-OPfp

Sequence: Fmoc-His(Boc)-OPfp


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₃₂H₂₆F₅N₃O₆
MW: 643.57 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237049
CAS Number: 109053-20-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR