Fmoc-Lys(FOR)-OH

Supplier: BACHEM AMERICAS INC

4014975.0005
B-1865.0005BAEA 722.61 CAD
B-1865.0005BA
Fmoc-Lys(FOR)-OH
Fmoc-Lys(FOR)-OH

Sequence: Fmoc-Lys(For)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₂H₂₄N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00237025
CAS Number: 201004-23-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR