N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine

Supplier: BACHEM AMERICAS INC

4010331.0005 4010331.0025
B-1395.0005BAEA 424.25 CAD
B-1395.0005BA B-1395.0025BA
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine

Sequence: Fmoc-p-nitro-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
p-Nitrophenylalanine (4-Nph, Phe(4-NO₂)) is incorporated into peptides as a chromophore. In FRET substrates, it acts as quencher. Bachem also offers resin-bound Fmoc-Nph (Fmoc-p-nitro-Phe-Wang resin, D-1135).

Formula: C₂₄H₂₀N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD00057810
CAS Number: 95753-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR