Boc-Glu-OPh

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Glu-phenyl ester

4029196.0001
A-4540.0001BAEA 258.14 CAD
A-4540.0001BA
Boc-Glu-OPh
Boc-Glu-OPh

Sequence: Boc-Glu-phenyl ester


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₆H₂₁NO₆
MW: 323.345 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057953
CAS Number: 59587-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR