Boc-D-Chg-OH

Supplier: BACHEM AMERICAS INC

4028496.0001 4028496.0005
A-4470.0001BAEA 309.81 CAD
A-4470.0001BA A-4470.0005BA
Boc-D-Chg-OH
Boc-D-Chg-OH

Sequence: Boc-cyclohexyl-D-Gly-OH


Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Chg derivatives.


Synonym(s): Boc-D-Chg-OH

Formula: C₁₃H₂₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00133629
CAS Number: 70491-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR