Boc-thionoleu-1-(6-nitro)benzotriazolide

Supplier: BACHEM AMERICAS INC

4030207.0001
A-4345.0001BAEA 901.53 CAD
A-4345.0001BA
Boc-thionoleu-1-(6-nitro)benzotriazolide
Boc-thionoleu-1-(6-nitro)benzotriazolide

Sequence: Boc-Thionoleu-1-(6-nitro)benzotriazolide


Activated building blocks for the synthesis of thioxopeptides. In the presence of the acid-labile thioamide (psi[CS-NH]) moiety, the Boc group can be removed with SnCl₄ in organic solvents.


Synonym(s): (S)-2-(Boc-amino)-4-methyl-pentanethioic O-acid-1-(6-nitro)benzotriazolide

Formula: C₁₇H₂₃N₅O₄S
Storage Temperature: Freezer
MDL Number: MFCD01318723
CAS Number: 214750-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR