Boc-n-Me-abz-OH

Supplier: BACHEM AMERICAS INC

Synonyms: 2-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid, 2-(N-Boc-N-methylamino)benzoic acid

4025576.0001 4025576.0005
A-3715.0001BAEA 309.81 CAD
A-3715.0001BA A-3715.0005BA
Boc-n-Me-abz-OH
Boc-n-Me-abz-OH

Sequence: Boc-N-Me-Abz-OH
Synonym(s): Boc-N-methyl-anthranilic acid#Boc-MANT

Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237567
CAS Number: 141871-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR