Boc-Dap(Fmoc)-OH

Supplier: BACHEM AMERICAS INC

4019047.0005
A-3580.0001BAEA 369.81 CAD
A-3580.0001BA A-3580.0005BA
Boc-Dap(Fmoc)-OH
Boc-Dap(Fmoc)-OH

Sequence: Boc-Dap(Fmoc)-OH


Abbreviated Dap, the abbreviation Dpr can be found in the literature as well. See also Albiziine, Nβ-carbamoyl-Dab, F-3670.

Formula: C₂₃H₂₆N₂O₆
MW: 426.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235897
CAS Number: 122235-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR