Boc-Glu-OMe

Supplier: BACHEM AMERICAS INC

4018552.0005 4018552.0025
A-3440.0005BAEA 464.57 CAD
A-3440.0005BA A-3440.0025BA
Boc-Glu-OMe
Boc-Glu-OMe

Sequence: Boc-Glu-OMe


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₁H₁₉NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00076931
CAS Number: 72086-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR