N-Boc-L-serine

Supplier: BACHEM AMERICAS INC

4001097.0025 4001097.0100
A-2275.0025BAEA 284.04 CAD
A-2275.0025BA A-2275.0100BA
N-Boc-L-serine
N-Boc-L-serine

Sequence: Boc-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.


Educt for the synthesis of Boc-Ser β-lactone, a useful synthon for obtaining stereochemically pure β-substituted α-amino acids. Building block for the incorporation of O-acyl isopeptide bonds during Fmoc-SPPS.

Formula: C₈H₁₅NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00037243
CAS Number: 3262-72-4
EINECS: 221-867-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR