Boc-N-Me-Val-OH

Supplier: BACHEM AMERICAS INC

4001829.0001 4001829.0005 4001829.0025
A-2100.0001BAEA 206.51 CAD
A-2100.0001BA A-2100.0005BA A-2100.0025BA
Boc-N-Me-Val-OH
Boc-N-Me-Val-OH

Sequence: Boc-N-Me-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
MeVal, αMeVal.


Rh-catalyzed carbene insertion into the N-H bond of the amide obtained from Boc-MeVal-OH yielded a Schmidt's phosphonate, educt for the synthesis of annulated diketopiperazines.

Formula: C₁₁H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00038760
CAS Number: 45170-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR