(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid

4009210.0005
A-1190.0005BAEA 645.24 CAD
A-1190.0005BA
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Sequence: Boc-Homophe-OH


Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.


Synonym(s): (S)-2-(Boc-amino)-4-phenyl-butyric acid

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076904
CAS Number: 100564-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR