Potassium iodide ACS

Supplier: VWR
Danger

0512-100G 0512-2.5KG 0512-500G
97061-828EA 123.27 CAD
97061-828 97061-830 97061-832
Potassium iodide ACS
Potassium iodide
Formula: Kl
Boiling Pt: 1330 °C (1013 hPa)
Melting Pt: 686 °C
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Barium 0.002%
Calcium = 0.002%
Chloride & Bromide = 0.01%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iodate = 0.0003%
Iron = 0.0003%
Loss on Drying = 0.2%
Magnesium = 0.001%
pH (5%, Water) @25°C 6 - 9.2
Phosphate = 0.001%
Purity = 99%
Sodium = 0.005%
Sulfate = 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR