Agarose, Molecular Biology Grade, IBI Scientific

Supplier: IBI Scientific
IB70045 IB70041 IB70046 IB70047 IB70042
87004-980EA 1661.26 CAD
87004-980 95043-304 97060-746 97060-748 490007-708
Agarose, Molecular Biology Grade, IBI Scientific
Electrophoresis Gels Agarose and Acrylamide
IBI Molecular Biology agarose products are highly refined and highly purified

Some features of IBI agarose include extraordinary mechanical resistance which insures safe and easy handling, exceptionally low absorption of staining reagents, excellent transparency which allows for high visibility, and greater thermal stability due to high hysteresis.

IBI agarose can be used for Nucleic Acid Electrophoresis, Analytical Electrophoresis, Preparative Electrophoresis, Blotting, and Protein Electrophoresis as in Radial Immunodiffusion.

Each lot is physically tested and verified forMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Ash content 0.4%
Clarity ≤3 at 1.0 % (NTU)
Electroendosmosis 0.05 - 0.13
Gelling temperature 36°C ± 1.5°C at 1.5 %
Gel strength ≥1200 at 1.0 % g/cm2 and ≥2500 at 1.5 % g/cm2
Moisture ≤7%
Sulphate 0.10%
Melting point 88°C ± 1.5°C at 1.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR