Anti-ASAH2 Rabbit Polyclonal Antibody

Supplier: Prosci
4743
89416-678EA 896.35 CAD
89416-678
Anti-ASAH2 Rabbit Polyclonal Antibody
Antibodies
ASAH2 Antibody: Sphingolipids are hydrolyzed by ceramidases to yield sphingosine and fatty acids. These ceramidases are classified according to the pH range that supports their optimal activity. ASAH2 is a neutral ceramidase and key regulator of sphingolipid signaling metabolites at the cell surface, catalyzing the hydrolysis of the N-acyl linkage of ceramide at an optimal pH of 6.5-8.5. ASAH2 is a type II integral membrane protein that can be cleaved to yield a soluble secreted protein and acts as a repressor of apoptosis both by reducing C16-ceramide, thereby preventing ceramide-inducedMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ASAH2
Antigen name N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2
Environmentally Preferable
Antigen synonyms ASAH2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID Q9NR71
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen ASAH2 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human ASAH2. The immunogen is located within the last 50 amino acids of ASAH2.
Form Liquid
Purification ASAH2 Antibody is affinity chromatography purified via peptide column.
Concentration 1 mg/mL
Storage buffer ASAH2 Antibody is supplied in PBS containing 0.02% sodium azide.
Storage temperature ASAH2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Shipping temperature 4 °C
Tested applications E, WB

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR