Anti-AMBRA1 Rabbit Polyclonal Antibody
Supplier: Prosci
4557
89416-550EA 896.35 CAD
89416-550
Anti-AMBRA1 Rabbit Polyclonal Antibody
Antibodies
Ambra1 Antibody: Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Beclin-1, a principal regulator of autophagosome formation, is in turn regulated by Ambra1. Ambra1 associates with Beclin-1 through a region near its center as determined by yeast two-hybrid assay. Null mutations in this gene in mice resulted in embryonicMore Product Information