Anti-TSC2 Rabbit Polyclonal Antibody
Supplier: Prosci
3509
89415-832EA 896.35 CAD
89415-832
Anti-TSC2 Rabbit Polyclonal Antibody
Antibodies
TSC2 Antibody: Tuberous sclerosis complex (TSC) is an autosomal dominant tumor syndrome caused by mutations in either of the TSC1 or TSC2 tumor suppressor genes. The products of these genes form a protein complex that indirectly decreases the signaling of the mammalian Target of Rapamycin (TOR), an evolutionarily conserved serine/threonine kinase that regulates cell growth and cell cycle through its ability to integrate signals from nutrient levels and growth factors. TOR activity is stimulated by Rheb, a member of the Ras superfamily of G-proteins, when the GTP/GDP ratio bound to Rheb isMore Product Information