Anti-TRIM5 alpha Rabbit Polyclonal Antibody
Supplier: Prosci
3251
89415-654EA 896.35 CAD
89415-654
Anti-TRIM5 alpha Rabbit Polyclonal Antibody
Antibodies
TRIM5 alpha Antibody: TRIM5 is a member of a broad family of otherwise unrelated proteins defined by the presence of a tripartite motif containing a RING domain, a B-box type 1, and a B-box type 2, followed by a coiled-coil region. TRIM5 has six alternately spliced isoforms, the longest of which is the alpha variant which also contains a carboxy-terminal B30.2 (SPRY) domain. Expression of TRIM5 alpha variants from humans, rhesus monkeys, and African green monkeys enabled resistance to infection by various retroviruses including HIV-1, albeit at differing efficiencies. All TRIM5 alphaMore Product Information