Anti-IKK alpha Rabbit Polyclonal Antibody
Supplier: Prosci
2025
89415-284EA 896.35 CAD
89415-284
Anti-IKK alpha Rabbit Polyclonal Antibody
Antibodies
IKK alpha Antibody: Nuclear factor kappa B (NF-kappa B) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa B mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFa, and bacteria product LPS. NF-kappa B is associated with I kappa B proteins in the cell cytoplasm, which inhibit NF-kappa B activity. The long-sought I kappa B kinase (IKK), which phosphorylates I kappa B, and mediates I kappa B degradation and NF-kappa B activation, was recentlyMore Product Information