Anti-DR3 Rabbit Polyclonal Antibody
Supplier: Prosci
1158
89415-254EA 896.35 CAD
89415-254
Anti-DR3 Rabbit Polyclonal Antibody
Antibodies
DR3 Antibody: Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel cell death receptor was recently identified by several groups independently and designated DR3, Wsl-1, Apo-3, TRAMP and LARD1-5. The ligand for this novel death receptor has been defined as TWEAK, also termed Apo3L. DR3 is highly expressed in the tissues enriched in lymphocytes including PBL,More Product Information