REDISHIP Protector® Premier® Laboratory Hoods and REDISHIP SpillStopper™ Work Surfaces, Labconco®
Supplier: Labconco
10040000010814 10050000010814 10060000010814 10080000010814
89260-050EA 18809.9 CAD
89260-050 89260-052 89260-054 89260-056
REDISHIP Protector® Premier® Laboratory Hoods and REDISHIP SpillStopper™ Work Surfaces, Labconco®
Fume Hoods and Enclosures Chemical Fume Hoods
Popular models of Protector® Premier® Laboratory Hoods and supporting matching work surfaces (sold separately) are available from VWR® stock for immediate shipment
REDISHIP SpillStopper™ Work Surfaces are dished to contain spills and feature a left rear corner cutout and oval cup sink with cover.
REDISHIP Protector® Premier® Fiberglass Hoods feature left and right-sided GFCI duplexes and Labconco's signature one-piece molded fiberglass reinforced polyester liner that provides superior corrosion and chemical resistance and durability. Each hood is factory prepared to accommodate up toMore Product Information