Acridine Orange 10-nonyl bromide (NAO) ≥95% (by HPLC), Ultrapure

Supplier: Enzo Life Sciences

ENZ-52306
89166-018EA 331.8 CAD
89166-018
Acridine Orange 10-nonyl bromide (NAO) ≥95% (by HPLC), Ultrapure
Acridine Orange 10-nonyl bromide (NAO)

Mitochondria dye


10-N-Nonyl acridine orange (NAO) is an acridine orange derivative and is generally used as a fluorescent marker of the inner mitochondrial membrane in whole cells. It is believed to bind to negatively charged phospholipids. NAO accumulation in the cell seems to be related to specific interactions with mitochondrial membrane proteins and/or lipids, such as cardiolipin, and is largely independent of mitochondrial membrane potential. With respect to apoptosis, the presence of mitochondrial membrane potential can be probed with Rhodamine123 while the structure and integrity of mitochondria can be assessed using 10-N-nonyl-acridine orange. Wavelength Maxima: Excitation 495nm, Emission 519nm

Formula: C₂₆H₃₈BrN₃
Storage Temperature: Freezer
MDL Number: MFCD00467605
CAS Number: 75168-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22fa5fb86b81969c3589d4c5c154738cebb8d6de&vid=b6126e837976f5393be1bc2887c502c633842454&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR