Zinquin free acid ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc

Supplier: Enzo Life Sciences

ENZ-52152
89165-982EA 331.8 CAD
89165-982
Zinquin free acid ≥98% (by HPLC), Ultrapure fluorescent indicator, for the determination of zinc
Zinquin

Zinc dye


Zinquin ethyl ester is a lipophilic, zinc-sensitive, fluorescent derivative of Zinquin that is able to penetrate cell membranes. Cleavage of the ethyl ester group via cytosolic esterases in living cells impedes its efflux across the plasma membrane. It is selective for Zn (II) in the presence of physiological concentrations of Ca (II) and Mg (II) ions. Fluorescence of Zinquin ethyl ester is enhanced upon binding Zn (II), but the probe does not demonstrate a shift or change in emission wavelength. The probe is UV-excitable and emits in the blue reqion of the spectrum. Zinquin ethyl ester has been used to monitor intracellular zinc fluxes associated with apoptosis. Wavelength Maxima: Excitation 368nm, Emission 490nm

Formula: C₁₉H₁₈N₂O₅S
MDL Number: MFCD03452711
CAS Number: 151606-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dea043f17ea3d7cd249289033514eaae0a832851&vid=10a4fe0c50dd0f551c804fa40170c6f0b3ac2307&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR