(K11-Only)Ubn-Ubiquitinylated Substrate
Supplier: Enzo Life Sciences
UW0620-0025
89165-188EA 496.12 CAD
89165-188
(K11-Only)Ubn-Ubiquitinylated Substrate
Proteins and Peptides
Protein components produced in E. coli. Synthesized enzymatically in vitro. (K11-only)Ubn-ubiquitinylated substrate protein was produced using K11-only mutant ubiquitin.