NOB ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST506-0050 BML-ST506-0250
89164-896EA 186.44 CAD
89164-896 89164-898
NOB ≥98% (by TLC)
NOB

PLA₂ substrate


  • Formulation: Off-white solid.


Substrate for phospholipases from snake venoms and human serum.


Soluble in DMSO (>25 mg/ml) or 100% ethanol (>25 mg/ml).


Synonym: 4-Nitro-3-(octanoyloxy)benzoic acid

Formula: C₁₅H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD00132973
CAS Number: 55894-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060ad0a3855f41a0620445d9057b6bbbe52c87cb&vid=9dce1d8d54f845352ff60b2dd6406ea564d6ff9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR