Human Thrombin (from Plasma)
Supplier: Enzo Life Sciences
BML-SE363-1000
89164-088EA 846.88 CAD
89164-088
Human Thrombin (from Plasma)
Enzymes
Isolated from human plasma as prothrombin, and activated with Factor Xa, Factor Va, phospholipids, and calcium to yield a-thrombin.