SC-51322 ≥98%

Supplier: Enzo Life Sciences

BML-RA112-0005 BML-RA112-0025
89162-500EA 172.22 CAD
89162-500 89162-502
SC-51322 ≥98%
SC-51322

Prostanoid receptor inhibitor


A potent and selective prostaglandin E2 antagonist (pA2=8.1). Displays in vivo analgesic activity in the mouse (ED50=0.9 mg/kg).

Formula: C₂₂H₂₀ClN₃O₄S
MDL Number: MFCD00916246
CAS Number: 146032-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06184601d37a37b7894a0b44989b4b3927b09aa6&vid=d7c710a3ffa59768f38a231a5b65b3101ae3efbc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR