Anti-PI31 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW9710-0025 PW9710-0100
89162-416EA 259.12 CAD
89162-416 89162-418
Anti-PI31 Rabbit Polyclonal Antibody
Antibodies
Recombinant PI31 has been shown to compete with the proteasome regulators PA28 and 19S PA700 for binding to 20S, indicating that PI31 binds the 20S α-rings and may function by hindering substrate access to the 20S catalytic channel.

  • For Western Blot

Recognizes human and mouse PI31.

Type: Primary
Antigen: PI31
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR