Anti-CYLD Rabbit Polyclonal Antibody
Supplier: Enzo Life Sciences
PW0760-0025 PW0760-0100
89161-946EA 224.36 CAD
89161-946 89161-948
Anti-CYLD Rabbit Polyclonal Antibody
Antibodies
The tumor suppressor CYLD is a deubiquitinylating enzyme (DUB) that antagonises NF-κB and JNK signaling by disassembly of Lys63-linked ubiquitin chains synthesised in response to cytokine stimulation. The DUB activity of CYLD is mediated by its C-terminal ubiquitin specific protease (USP) domain, pathogenic truncations and mutations of which are associated with the hypertrophic skin tumor cylindromatosis through disruption of the USP domain and subsequent loss of CYLD catalytic activity.
  • For Western Blot
Recognizes human CYLD.
Type: Primary
Antigen:More Product Information