Anti-Ubiquitin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW0600-0025 PW0600-0100
89161-922EA 237 CAD
89161-922 89161-924
Anti-Ubiquitin Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2a

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes K63-linked polyubiquitin in wide range of species. Does not cross-react with any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitinylated species.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR