Z-Leu-Leu-Leu-B(OH)2
Supplier: Enzo Life Sciences
BML-PI109-0100
89161-588EA 529.3 CAD
89161-588
Z-Leu-Leu-Leu-B(OH)2
Proteins and Peptides
Proteasome inhibitor
A potent cell-permeable proteasome inhibitor (Ki=0.023 nM).