Calpeptin ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-PI101-0010 BML-PI101-0050
89161-562EA 249.64 CAD
89161-562 89161-564
Calpeptin ≥98% (by TLC)
Calpeptin

Calpain and cathepsin inhibitor


Membrane permeable inhibitor of calpain I and II, cathepsin L and cathepsin K (IC50=0.11 nM). Promotes neurite elongation in differentiating PC12 cells. Prevents Ca2+-ionophore induced degradation of actin binding protein and P235 in platelets. Inhibits myosin light chain phosphorylation in platelets stimulated by collagen, ionomycin (Prod. No. ALX-450-006) or thrombin. Inhibits LPS induced iNOS transcription and NF-kappaB activation, possibly via proteasome inhibition.

Formula: C₂₀H₃₀N₂O₄
MDL Number: MFCD00155623
CAS Number: 117591-20-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceabbeba93761b23f18d336486a2d663a0669c83&vid=80f83cb3ef6a1b40ca107c3ee5ec91773a265c30&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR