6-Ketoprostaglandin-F1α ≥98% (by TLC)

Supplier: Enzo Life Sciences
Warning

BML-PG017-0001
89161-474EA 252.8 CAD
89161-474
6-Ketoprostaglandin-F1α ≥98% (by TLC)
6-Ketoprostaglandin-F1alpha

Biosynthesis via hydrolysis of prostacyclin. A reliable parameter for measurement of prostacyclin. Ultra-pure.


  • Formulation: White to off-white solid.


Soluble in ethanol (25 mg/ml).

Formula: C₂₀H₃₄O₆
Storage Temperature: Freezer
MDL Number: MFCD00135222
CAS Number: 58962-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72862d64466bc623b427885fb0d72c7807fd82f&vid=85a06c7f71f88afdf3303c2b41bbfd7601a6c0bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (TLC)
Appearance White to off-white solid

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR