Ac-REQD-pNA
Supplier: Enzo Life Sciences
BML-P437-0005
89161-246EA 254.38 CAD
89161-246
Ac-REQD-pNA
Proteins and Peptides
Caspase substrate
Colorimetric substrate for caspase-6 (Km=30µM; Kcat/Km=168’000 M-1sec-1) and related cysteine proteases. Sequence is based on the lamin A site of cleavage. Cleavage of the substrate by a caspase releases pNA, which can be detected by its absorbance at 405nm (ε=10,500 M-1cm-1).