Leu-Pna (Chromogenic Substrate)
Supplier: Enzo Life Sciences
BML-P230-0100
89161-114EA 213.3 CAD
89161-114
Leu-Pna (Chromogenic Substrate)
Proteins and Peptides
Aminopeptidase substrate
Substrate for aminopeptidases such as aminopeptidase N (APN) cystinyl aminopeptidase, and leucine aminopeptidase (LAP). This substrate can be used for in vitro assays with purified enzyme, or with cell extracts. Also available: calibration standard pNA (BML-KI145).