Acitretin ≥98%

Supplier: Enzo Life Sciences

Synonyms: (2E,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid

BML-GR108-0010
89158-764EA 252.8 CAD
89158-764
Acitretin ≥98%
Acitretin

Retinoic acid receptor ligand.


  • Formulation: Yellow solid.


Acitretin is a synthetic aromatic analog of all trans retinoic acid with increased stability in vivo. It is the active free acid form of the antipsoriatic drug etretinate. Displays antiproliferative effect and induces apoptosis in kaposi's sarcoma cells.


Soluble in DMSO.

Formula: C₂₁H₂₆O₃
MDL Number: MFCD00866632
CAS Number: 55079-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ed79ca18e560df0153ddd7c281e9293bef6b1585&vid=3ce27ab19b5015f55312bfa36a992b93f7d7906b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR