BML-277 Chk2 inhibitor ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-EI388-0010
89158-242EA 237 CAD
89158-242 89158-244
BML-277 Chk2 inhibitor ≥98% (by TLC)
BML-277 Chk2 inhibitor

BML-277 is a highly selective inhibitor of Chk2 (IC50=15nM). It displays potent radioprotective activity and prevents apoptosis of human T cells subjected to ionizing radiation (EC50= 3-7.6µM). Useful tool for dissecting the role of Chk2 in cellular signaling.


  • Chk inhibitor


Off-white solid.


Soluble in DMSO (20mg/ml) or ethanol (20mg/ml).

Formula: C₂₀H₁₄ClN₃O₂
Storage Temperature: Freezer
MDL Number: MFCD08276917
CAS Number: 516480-79-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89f71c62aebc21ccf249d5ed7b846b31807cd863&vid=05653ac1b243ce076c3a2fe20901d3bfba2dfadc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR