Chelerythrine chloride ≥98%

Supplier: Enzo Life Sciences

BML-EI225-0005 BML-EI225-0025
89157-808EA 188.02 CAD
89157-808 89157-810
Chelerythrine chloride ≥98%
Chelerythrine chloride

PKC inhibitor


Reported to be a potent and selective inhibitor of protein kinase C (IC50=0.66µM). Although chelerythrine has numerous documented biological effects, studies have raised serious doubts about the effectiveness of chelerythrine as a PKC inhibitor.

Formula: C₂₁H₁₈ClNO₄
MDL Number: MFCD00060717
CAS Number: 3895-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96b18c46455e9fd49dfd3afb0daab8a59726d741&vid=70cf371de9db9a542c51a74c7c28f7efcbe75bd4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR