Proadifen hydrochloride ≥95% (by TLC)

Supplier: Enzo Life Sciences

BML-EI105-1000
89157-564EA 176 CAD
89157-564 89157-566
Proadifen hydrochloride ≥95% (by TLC)
Proadifen hydrochloride

Cytochrome P450 inhibitor


Cytochrome P-450 inhibitor. Inhibits cytochrome P-450-dependent arachidonate metabolism (90% at 50 µM). Calcium antagonist. Blocks glibenclamide-sensitive K+ channels (IC50=4.4mM). Inhibits platelet thromboxane synthesis and stimulates endothelial cell prostacyclin production. Potentiates the effects of many drugs in vivo. 

Formula: C₂₃H₃₁NO₂·HCl
MDL Number: MFCD00055151
CAS Number: 62-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9ede98ffc21613f95fd794ffc72a32afea3b9a6&vid=b63a7e2323eb2e708240ee6692d7ca3886b3f22b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR