Forskolin ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-CN100-0010 BML-CN100-0100
89156-986EA 248.06 CAD
89156-986 89156-988
Forskolin ≥99% (by TLC)
Forskolin

Adenylate cyclase activator


Activates adenylate cyclase by directly interacting with the catalytic unit of the enzyme leading to an increase in the intracellular concentration of cAMP. Widely used tool for the investigation of the role of cAMP as a second messenger with a broad range of potential therapeutic applications. Inotropic agent and vasodilator. Induces platelet aggregation. Inhibits ion channels by a mechanism that does not involve cAMP. Non-competitive inhibitor of nicotinic acetylcholine receptors.


Isolated from Coleus forskohlii.

Formula: C₂₂H₃₄O₇
MDL Number: MFCD00082317
CAS Number: 66575-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbaaa80dc33de2287e1d9a98b63daffe6e1bcb19&vid=29bdd5e4828e1ba56f5c573017c8f2b3840b6fad&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR