4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)

Supplier: Enzo Life Sciences

BML-C104-0050
89156-694EA 115.34 CAD
89156-694
4-Diphenylacetoxy-N-methylpiperidine methiodide ≥98% (by HPLC)
4-Diphenylacetoxy-N-methylpiperidine methiodide

Acetylcholine receptor ligand.


  • Formulation: White solid.


M₃ muscarinic receptor antagonist.


Soluble in DMSO and warm water.

Formula: C₂₁H₂₆INO₂
Storage Temperature: Ambient
MDL Number: MFCD00078564
CAS Number: 1952-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0bc9965638f9641add2f779652c775443cd955e&vid=b8bc7be8e562a8e6cd43ee1aeee73fa34be80c1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR