1,5-Isoquinolinediol ≥98%

Supplier: Enzo Life Sciences

BML-AP102-0020 BML-AP102-0100
89156-568EA 121.66 CAD
89156-568 89156-570
1,5-Isoquinolinediol ≥98%
1,5-Isoquinolinediol

PARP inhibitor


Potent inhibitor of inducible nitric oxide synthase (iNOS; NOS II) in mouse macrophages. Potent and selective PARP inhibitor. Inhibits nitric oxide (SIN-1) induced PARP activation in rat hepatocytes (at 20µM). Neuroprotectant.

Formula: C₉H₇NO₂
MDL Number: MFCD00006900
CAS Number: 5154-02-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2444fe4436b20a42eab7ca01ded1b692de7f510a&vid=612616cba818f4d81e41c5a58b180ca90e004de7&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR