Anti-CCL22 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-469-C250
89154-348EA 1564.2 CAD
89154-348
Anti-CCL22 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human CCL22.

Type: Primary
Antigen: CCL22
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CCL22
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Reactivity Human
Host Rat
Isotype IgG
Western blot Yes
ELISA Yes
Form Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives.
Purification Protein G-affinity purified
Concentration ~1mg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR