Anti-PMAIP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-408-C100
89154-216EA 796.32 CAD
89154-216
Anti-PMAIP1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human and mouse Noxa. Detects a band of ~11kDa by Western blot.

Type: Primary
Antigen: PMAIP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR