Anti-VASP Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-403-C100
89154-202EA 1232.4 CAD
89154-202
Anti-VASP Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human and mouse Ser157-phosphorylated VASP. Does not cross-react with the non-phosphorylated form of VASP.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: VASP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR