Anti-IL6 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-335-C100
89154-058EA 538.78 CAD
89154-058
Anti-IL6 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG1

  • For ELISA, Flow Cytometry
  • Endotoxin content: <0.004EU/µg

Recognizes mouse IL-6.

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR