Anti-Endothelin-1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
803-001-R100
89153-370EA 1188.16 CAD
89153-370
Anti-Endothelin-1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Western Blot, RIA

Recognizes human, rat and sheep ET-1 (epitope aa 8-16). Cross-reacts slightly with related peptides.

Type: Primary
Antigen: ENDOTHELIN-1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR