Zearalenone ≥99% (by HPLC, TLC)

Supplier: Enzo Life Sciences
Danger

630-105-M010
89153-000EA 251.22 CAD
89153-000 89153-002
Zearalenone ≥99% (by HPLC, TLC)
Zearalenone

Mycotoxin.


  • Formulation: White to off-white powder.


Zearalenone (ZEN) is an estrogenic mycotoxin in animals and a phytohormone in plants. Inducer of sister chromatid exchange and chromosomal aberration. Acts as a protonophoric uncoupler in plant mitochondria.


Soluble in methanol (5 mg/mL).


Synonym: Benzoxacyclotetradecin-1,7(8H)-dione; ZEN.

Formula: C₁₈H₂₂O₅
Storage Temperature: Freezer
CAS Number: 17924-92-4
EINECS: 241-864-0
UN: 2811
ADR: 6.1,II
Merck Index: 14,10116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9b2813118c79b195b7723cce63fc7cf6e9e665d&vid=371a0a9b9df58af47f01832afa466ff75e08d657&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Isolated from Giberella zeae
Purity ≥99% (HPLC, TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR