AIDA ≥96%

Supplier: Enzo Life Sciences

Synonyms: (±)-1-Aminoindane-1,5-dicarboxylic acid

550-101-M025
89152-154EA 322.32 CAD
89152-154
AIDA ≥96%
AIDA

mGluR agonist.


Potent and selective ligand for group I mGluRs. Shows no effect on group II or group III mGluRs. Has no effect on ionotropic glutamate receptors.


Soluble in water.


Synonyms: (±)-1-Aminoindane-1,5-dicarboxylic acid, UPF 523

Formula: C₁₁H₁₁NO₄
Storage Temperature: Refrigerator
CAS Number: 168560-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7aa21ec58b5bdf9187f867ce25698dfc75a49f86&vid=b703b68ec908b86395920af1742f327d8b9567b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR