6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)

Supplier: Enzo Life Sciences

550-042-M005 550-042-M010 550-042-M025 550-042-M050
89152-044EA 109.02 CAD
89152-044 89152-046 89152-048 89152-050
6-Cyano-7-nitroquinoxaline-2,3-dione ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione

AMPA/kainate receptor antagonist


  • Formulation: Off-white to yellow powder.


Potent, competitive kainate/quisqualate (non-NMDA) receptor antagonist. For the more soluble form see CNQX . disodium salt (Prod. No. ALX-550-283).

Formula: C₉H₄N₄O₄
MDL Number: MFCD00069232
CAS Number: 115066-14-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0291c68fd70638abfb66ca8fc4456825dc513f9&vid=447e2c05219280bbb48c75c966981fbddf232637&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR